Advanced Signal Generator
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SE1082
470229-040EA 610.95 USD
470229-040
Advanced Signal Generator
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
High power and versatility.
  • Low distortion with four decade ranges
  • Produces square, sine, triangle, and TTL waveforms
  • Built-in speaker
  • Equipped with provisions for AM and FM modulation
  • A weak external signal can also be amplified through the unit
Ideal for general-purpose applications in a student laboratory, particularly regarding wave phenomena. Unit has an output range of 10Hz to 110KHz, in four decade ranges.