Soldering Gun
Supplier: KIDDER MFG. CO. LTD
80-4859-00
470228-924EA 67 USD
470228-924
Soldering Gun
Soldering Supplies Soldering Irons
More comfortable style to hold than traditional design.
  • Pistol grip
This soldering gun is 100 watts. 40 gm solder tube included.
Certifications: CSA approved.