CoolCaddy™ Cold WorkStation
Supplier: MTC BIO, INC.
R4015
470228-446EA 170.2 USD
470228-446
CoolCaddy™ Cold WorkStation
Coolers Benchtop Coolers
Keeps your samples cool while you experiment.
  • Maintains 1 to 4 °C for up to 5 hours
  • Keeps samples cool but not frozen
  • Eliminates the need for crushed ice
  • Accomodates most small tube types
Keeps your reagents and samples cool on the lab bench without the use of crushed ice.
Simply freeze overnight and its ready to hold 1 to 4 °C for up to 5 hours, keeping your Microtubes, Cryovials, PCR Tubes/Plate and Reagent Reservoirs cold (but not frozen) while you work.