BoneClones® Human Male Cranium with Hammer Blows
Supplier: BONE CLONES HOLDINGS
BC-217
470228-138EA 416.65 USD
470228-138
BoneClones® Human Male Cranium with Hammer Blows
Models Human Biology Models
An excellent model of blunt force trauma.
  • Shows two rounded wounds
  • Smooth edges indicate healing
  • Excellend model of blunt force trauma
This blunt force trauma skull illustrates two wounds that are rounded and depressed, consistent with hammer injury. The smooth, rounded edges are indicative of healing, and therefore indicate some period (likely months or years) of survival.
Ordering information: The osteological evaluation report (PDF) will be sent with the purchase of this specimen.