MicroPen Microscope
Supplier: CARSON OPTICAL
MP-300
470227-898EA 19 USD
470227-898
MicroPen Microscope
Magnifiers
A microscope the size of a pen! Use this in an industrial setting, the classroom, or the back yard.
  • 24X–53X magnification
  • 3G3 button cell batteries
  • Industrial use
  • Classroom use
  • Built-in LED
  • Focus and zoom adjustments