MiniScout Binoculars
Supplier: CARSON OPTICAL
JD-718
470227-884EA 45 USD
470227-884
MiniScout Binoculars
Monoculars and Binoculars
Ultra-compact binocular with 7X and a center focus knob.
  • Fully coated lenses
  • Lightweight at 4.5 oz.
  • Independent right diopter
Ordering information: Includes pouch, neck strap, and lens cloth.