Cometron 7X50
Supplier: Celestron International
71198
470227-850EA 45.89 USD
470227-850
Cometron 7X50
Monoculars and Binoculars
Ideal binocular for comet watching, stargazing, and viewing craters on the Moon.
  • BK7 Prism Glass
  • 7X magnification
  • 50 mm objective lens diameter
  • Wide neck strap and carrying case included
Wide field of view reveals a larger portion of the night sky, allowing you to view more of the comet's impressive tail. Large 50mm objective lenses have tremendous light-gathering ability, ideal for astronomical use. Multi-coated optics dramatically increase light transmission for brighter images with increased contrast and resolution. Large exit pupil provides maximum light during nighttime and low-light viewing sessions. Strong and durable alumnium housing.