Hall Effect Apparatus
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SEB01
470121-160EA 1590.7 USD
470121-160 470227-534
Hall Effect Apparatus
Educational Materials Physics Educational Materials Quantum Mechanics Learning Activities
This apparatus determines hall voltage and coefficient.
  • Heavy-Duty Electromagnet
  • Gauss Meter with a Thin Probe
  • Manual
  • Semiconductor Ge Sample
  • Digital Multimeter
The most distinguishing feature allows the student to individually mount a Ge crystal.
Students can also determine the type of charge carrier, the charge density of carriers, and the resistivity of a given sample.
Ordering information: The unit includes a Gauss meter with probes; digital multimeter; semiconductor 'Ge' sample, p type; and Hall effect apparatus. A 12V power supply is required but not provided.