Ward's® Essentials Green Glucose Production Kit
Supplier: Avantor
470227-320KT 99.99 USD
470227-320
Ward's® Essentials Green Glucose Production Kit
Educational Classroom Kits and Activities
Measure photosynthesis and respiration using algae beads.
  • Hands on lab activity
  • Kit aligned to Next Generation Science Standards MS-LS1-6 and MS-ETS1-1
  • Instructions and materials included for 15 groups
  • Middle school
A simple organism, algae beads, are used to determine the rates of photosynthesis and respiration in a colorimetric assay. Using a pH indicator, test the effects of light on these important cellular processes.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, email or online.
Caution: WARNING: CHOKING HAZARD–Product contains small parts, small balls, or marbles. Not for children under 3 yrs.