Deluxe Learn to Solder Kit
Supplier: Elenco Electronics
SK175
470227-034KT 47.5 USD
470227-034
Deluxe Learn to Solder Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Soldering set for the advanced user.
  • Learn how to solder properly
Learn how to solder properly using a variable temperature solder station providing a temperature range of 350° to 900°F. It will provide the correct temperature for the broad range of electronic components that are present in today's electronic equipment.
SOLDERING REQUIRED!
Ordering information: This kit contains: variable temperature station with conical tip; wedge tip; solder practice kit; side cutters; and solder ease kit. All packed together in a handy box with handle for carrying.