Digital Temperature Controlled Solder Station
Supplier: Elenco Electronics
SL30A
470227-030EA 183 USD
470227-030
Digital Temperature Controlled Solder Station
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Professional Soldering Station 48W (160-480 C or 320-900 F) with a 42 inch long cable.
  • Heater Power for a 48 watt Soldering Iron
  • Iron Voltage is 24 VAC (station runs on 110-130 VAC 60Hz)
The digital display is switchable to read out in C or F.