NeuLog Rechargeable Battery Module
Supplier: Wards
BAT-200 BAT202
470137-440EA 54.09 USD
470137-440 470226-926
NeuLog Rechargeable Battery Module
Data Loggers
Make your Neulog probes portable with the battery module.
  • Battery module supplies power to a sensor, or chain of sensors, operating in the offline mode, and to sensors connected to an RF communication module
The battery module can be recharged by connecting it to the PC’s USB socket via the USB module’s USB cable. The battery module has a LED and a pushbutton switch. The LED indicates whether the battery is charged enough or not when pressing the pushbutton.
Caution: For educational use only.