Electric Whirl
Supplier: Wards
PH0904
470226-556EA 44.1 USD
470226-556
Electric Whirl
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate rotary motion from a charge.
  • For demonstrating the effect of electric discharge from points
The apparatus has a 95 mm diameter, Three-pointed star wheel with all the points curved in one direction. The wheel rotates freely on a pivot point mounted on a short stand and the stand is provided with a 4 mm connection socket.