Denoyer-Geppert® Spinal Palpation Trainer
Supplier: Denoyer-Geppert Science
SP672
470225-910EA 267.35 USD
470225-910
Denoyer-Geppert® Spinal Palpation Trainer
Models Human Biology Models
The palpation training kit includes two levels of tissue simulation to inform the varying levels of soft tissue that exists throughout the general population and spine.
  • Practice palpation through 2 thicknesses of tissue.
This new spine palpation trainer simulates the soft tissue (muscle, skin, and fat) that overlies the vertebral column, allowing the user to improve their tactile and kinesthetic skills at knowing which vertebral level and which spinous process is being palpated. Perfect for practitioners and students of massage, physical therapy, chiropractic's, osteopathic, or orthopedics.