Innovating Science® Properties of Aspirin
Supplier: Ward's Science
IS9354 IS9354-SGL
470225-580EA 87.1 USD
470225-580 470231-574
Innovating Science® Properties of Aspirin
Educational Materials Chemistry Educational Materials Consumer Chemistry Learning Activities
What is Aspirin made of?
  • Classic AP chemistry experiment
In a reaction vessel salicylic acid, acetic anhydride and sulfuric acid are mixed. The exothermic reaction will cause the temperature to increase to 70 to 80 °C. Once the reaction is complete the vessel is cooled in an ice bath and the acetylsalicylic acid crystallizes out. This material is re-crystallized in toluene to purify the product. The product identity is confirmed by melting point. The mass of the re-crystallized product is used to determine the yield of the synthesis. In this experiment, we will perform the synthesis of acetylsalicylic acid (aspirin), purification by re-crystallization and use melting point to confirm identity.
Delivery information: Instructor's Manual and SDS included.