Ward's® Essentials Properties of Polymers Lab Kit
Supplier: Ward's Science
AD-WE2505-KIT
470225-524EA 68.99 USD
470225-524
Ward's® Essentials Properties of Polymers Lab Kit
Educational Classroom Kits and Activities
Learn about and create three types of polymers and explore the differences.
  • Hands on lab activity
  • Kit aligned to next generation standards MS-PS1-2, MS-PS1-3
  • Instructions and materials included for 15 groups
  • Middle school
Students will create polymer worms, polymer slime, and polymer snow as they explore differences between thermoplastic and thermoset polymers. Students will also investigate properties of various crosslinked, superabsorbent polymers.