EduPrimer™ VNTR DNA Profiling Kits
Supplier: GENOSENSOR CORPORATION
3003
470225-434EA 223 USD
470225-434
EduPrimer™ VNTR DNA Profiling Kits
Educational Materials Biology Educational Materials PCR Learning Activities
The EduPrimer™ DNA profiling kit is designed to introduce students to Variable Number Tandem Repeats (VNTRs).
  • Highly variable results between students
  • Fast and simple protocol for easy integration into biotechnology or forensics lessons
  • Introduce students to DNA isolation, PCR and DNA electrophoresis
The EduPrimer™ DNA Profiling Kit is designed to introduce students to the principles and applications of PCR, focusing on the real-life use of DNA Profiling in forensics and disease research.
The Penta E microsattelite region, a Variable Number Tandem Repeat (NVTR targeted with this kit makes it an excellent example of genetic elements that can be used for identification. Compare results between your students, and between classes.
Ordering information: This kit includes the components needed to isolate DNA from students, as well as the PCR components needed to amplify DNA. Kits come with directions to download instructional material and procedures. Kit includes DNA isolation components (lysis buffer/neutralizer), PCR reaction components including master mix (primers and enzymes) and DNA ladder.
Equipment needed but not included: Microcentrifuge, PCR thermal cycler, micropipettes and tips, PCR tubes, microcentrifuge tubes, heat block or water bath, TAE buffer, DNA Electrophoresis equipment/reagents including transilluminator/light box for analysis.
Delivery information: Kits include perishable materials, please provide a delivery date at check-out.
Packaging: Kits contain reagents only.