Chrome Steel Bar Magnet
Supplier: American Scientific
3101-33-B 3101-33
470225-330PR 26.6 USD
470225-330 470225-382
Chrome Steel Bar Magnet
Magnets
For demonstrations, plotting magnetic fields.
  • Sturdy zinc plated magnets
  • Comes in marked and unmarked styles
Bar Magnet, made from magnetic steel strips, zinc plated. Supplied in pairs with cardboard separator and plated soft iron keepers.