Ward's® Essentials Catapult Kit
Supplier: Wards
VWRGP00007
470225-378KT 45.29 USD
470225-378
Ward's® Essentials Catapult Kit
Educational Classroom Kits and Activities
Play with the physics of a catapult to learn about levers, force, acceleration and much more!
  • Hands on lab activity
  • Kit aligned to Next Generation Science Standards MS-PS2-2
  • Instructions and materials included for 1 group
  • Middle school
Explore force, acceleration, Hooke's law, and parabolic trajectories by building and testing a fully-functional catapult. Contains catapult base, sides, lever arm, rubber bands, screws, dowels, hook, and ping-pong ball.