Sine Wave Signal Generator
Supplier: American Scientific
8410-00
470225-332EA 206.9 USD
470225-332
Sine Wave Signal Generator
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Create waves and signals quickly and easily.
  • This sine wave generator is a great economical tool for generating waves with speakers or wave drivers
It allows both the frequency (1-4,000 Hz) and amplitude of the sine wave output to be varied. The digital display features a 0.1 Hz resolution.