3B Scientific® Mitosis Model
Supplier: American 3B Scientific
1013868
470224-956EA 787.45 USD
470224-956
3B Scientific® Mitosis Model
Models Molecular Biology Models
The three-dimensional relief model shows 9 phases of mitosis on the basis of a typical mammal cell.
  • Chromosomes colored according to modified AZAN staining colors
  • Cell components are color-coded in accordance with educational aspects
  • Attaching magnets on the rear
  • Storage system, free-standing or hanging up
  • Supplied with detailed description and copying templates
  • 3 year manufacturers warranty
1. Interphase, 2. Prophase, 3. Early prometaphase, 4. Later prometaphase, 5. Metaphase, 6. Early anaphase, 7. Later anaphase, 8. Telophase, 9. Cytokinesis.