EclipSmart Roof Solar Binoculars, 10×25
Supplier: Celestron International
71237
470224-449EA 49.89 USD
470224-449
EclipSmart Roof Solar Binoculars, 10×25
Monoculars and Binoculars
Easy and safe binoculars to observe the sun during an eclipse or for general solar viewing on the go.
  • Premium EclipSmart solar binoculars featuring solar safe filter technology for guaranteed safe observation of the sun
  • Dual lens non-removable glass solar filters fully meet ISO12312-2 - safe viewing of the sun when used in accordance with the included instructions
  • The generous field of view allows users the ability to easily locate and view the entire solar disk while observing
  • Observe the natural details of the sun, including daily sunspot activity
  • Durable and rugged for protection during use and storage provided by the rubber armored aluminum body
Whether eclipse viewing or general solar viewing, this line of products is the one for you. Solar safe technology. EclipSmart solar products feature solar safe filter technology providing the ultimate protection from harmful solar radiation, including both IR and UV light, and filters 99.999% of visible light. Celestron solar safe filter technology is GUARANTEED SAFE for direct solar observation and has been independently tested by SAI global assurance services.
Small and mighty: The celestron EclipSmart 10×25 mm solar binocular is compact in size making it the perfect outdoor companion. The size makes it easy to carry from one viewing location to another and can even fit within a coat pocket! these binoculars also provide a generous field of view allowing for a full image of the solar disk to be seen.