Miniature LED Lights
Supplier: Skywill Far East Industries
OPDT-W4DK86A1B
470163-914PK 23.05 USD
470163-914 470224-034
Miniature LED Lights
Lights LED Lights, Lamps and Bulbs
These lights are great to use with a variety of electricity and science projects.
  • Your batteries will last longer: eco-friendly LEDs draw only 25 mA
  • Bulbs light only when current is in correct direction
  • Size E10 screw base
  • Pack of 10
Perfect for student investigations of DC circuits. Not recommended to exceed 3 V. The bulbs light only when the current is in the correct direction.
Packaging: One package has 10 bulbs.