National D-ELS-4 Zoom Stereomicroscope
Supplier: National Optical
D-ELS-4
470222-460EA 698.99 USD
470222-460
National D-ELS-4 Zoom Stereomicroscope
Microscopes
The D-ELS-4 model is a new stereo-zoom microscope perfect for high school students.
  • Zoom: 1:6
  • Magnification: 0.7x–4.5x
  • Dual LED Energy-Efficient Illumination Lasts up to 50,000 Hours
  • Three-Year Limited Manufacturer's Warranty
  • Designed for High School Classes
It allows students to view objects at .7x magnification then zoom to 45x magnification for an exciting, close-up view.
The microscope has energy-efficient LED illumination that lasts up to 50,000 hours.
Ordering information: This product includes a manufacturer's warranty.