Ohaus® Frontier™ 5000 Series Mini Centrifuge
Supplier: OHAUS CORP.
30134157
470222-278EA 293.9 USD
470222-278
Ohaus® Frontier™ 5000 Series Mini Centrifuge
Centrifuges Microcentrifuges
The Ohaus Frontier 5306 is a small but powerful minicentrifuge for simple and quick liquid separation.
  • Centrifuge Accommodates Multiple Sizes of Micro Tubes
  • Quiet, Safe, Easy Operation
  • Perfect for Quick Spin-Down Applications
  • Tube Rotor: 1.5 mL
  • Centrifuge Can Accommodate 32 Individual PCR tubes
With an eight-place, 1.5 mL tube rotor, Frontier 5306 can accommodate 0.2 mL eight-strip tubes or 32 individual PCR tubes. With the included adapter, 0.2 mL and 0.5 mL tubes can also be accommodated in the eight-place tube rotor.
Operation is quiet, easy, and safe, making this centrifuge perfect for spin-down applications.