Kestrel 3500 Fire Weather Meter
Supplier: NIELSEN-KELLERMAN CO
0835FWORA
470222-121EA 371.65 USD
470222-121
Kestrel 3500 Fire Weather Meter
Weather Stations
The Kestrel 3500 has the most functions of any Kestrel Meter in the 3000 series. It measures wind speed, relative humidity, temperature, wet bulb temperature (psychrometric), altitude, pressure trend, barometric pressure, heat stress index, dew point temperature, wind chill.
  • Easy to use system scrolls through 9 different environmental conditions
  • Waterproof and floats
  • Large easy to read display
Slip on protective cover included.