Metal Backed Student Thermometer, United Scientific Supplies
Supplier: United Scientific Supplies
THMC02
470222-092EA 5 USD
470222-092
Metal Backed Student Thermometer, United Scientific Supplies
Thermometers Analog Thermometers
Great for classroom use.
  • Glass thermometer mounted on aluminum back
  • Filled with kerosene solution
  • Celsius only
Glass thermometer mounted on aluminum back. Thermometers are filled with approximately 3 grams of kerosene mixed with red color.