Simple Anemometer
Supplier: United Scientific Supplies
ANMM01
470221-652EA 67.35 USD
470221-652
Simple Anemometer
Anemometers
This simple tool measures wind speed and direction.
  • A built-in compass is provided
The wind speed range is 0 to 17 meters per second (+/-10%). The direction indicator is divided into 16 compass points.