Wet and Dry Bulb Wall Thermometer
Supplier: United Scientific Supplies
THWD01
470221-650EA 9.25 USD
470221-650
Wet and Dry Bulb Wall Thermometer
Thermometers Analog Thermometers
Measure of temperature and humidity.
  • Includes humidity table in C and F
Graduated in Celsius -20° to +50° (1°C) and Fahrenheit 0° to +120° (2°F).