Rudiger® Pelvic Floor Muscles And Flexible Pelvis
Supplier: RUDIGER ANATOMIE GMBH
A276
470221-158EA 387.65 USD
470221-158
Rudiger® Pelvic Floor Muscles And Flexible Pelvis
Educational Materials Biology Educational Materials Human Biology Learning Activities
High quality model of the female pelvis with pelvic floor muscles.
  • Fully flexible pelvis
  • Shows interactions of pelvis and pelvic floor
  • 5 year manufacturers warranty
Not dissectible, but with flexible assembly to demonstrate pelvic movement. All important muscles are shown. Sliding movement and rotational movements are possible between pelvis and sacrum, Comes with key and numbering