Mystery Density Activity Set
Supplier: Wards
PH0113MDS
470220-792EA 73.1 USD
470220-792
Mystery Density Activity Set
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Demonstrates the relationship between mass and volume.
  • Set of 12 bars of different length but the same diameter
  • Comes in an attractive wooden holder
As students plot the first few points it appears there is no relationship between the mass and volume. However, as more data is graphed, it becomes obvious that two different linear relationships exist. They are actually made of two different materials.