Eisco® Folding Quadrat
Supplier: Wards
BI0504
470220-788EA 28.05 USD
470220-788
Eisco® Folding Quadrat
Samplers Solid Samplers
Hands-on field sampling tool.
  • Square sampling quadrat
  • Folds flat when not in use
  • Used for sampling plants and slow moving organisms in the field
  • Aligned with GLOBE protocols
For population counts and other measurements in the field. Consists of four white coated steel rods 0.5 m long with markers at 0.25 m, which simply bolt together to form a quadrant frame.