Monocot Stem Structure
Supplier: United Scientific Supplies
MBMS01
470220-636EA 103.25 USD
470220-636
Monocot Stem Structure
Educational Materials Biology Educational Materials Plant Learning Activities
Students can use this structure to examine a corn stem in cross and longitudinal sections.
  • Model Displays Basic Structure of Monocot Stem
  • Large Size for Easy Examination
  • Detailed Key Included
  • Durable Construction
  • Injection-Molded Model
This model is patterned on a corn stem to show the general anatomy of a monocot in both longitudinal and cross sections.  It reveals features such as the cuticle layer, epidermis, closed vascular bundles, and parenchyma cells.
This injection-molded model is more durable than the more common fiberglass construction. 
Ordering information: This structure includes a detailed key.