Lens and Mirror Set
Supplier: Wards
PH0201601
470220-586EA 59.05 USD
470220-586
Lens and Mirror Set
Educational Materials Physics Educational Materials Optics Learning Activities
Perfect compliment for a ray box.
  • Universal mirror
  • Frosted lenses
Optics lens/mirror Kit, includes 1, Acrylic Double Convex frosted lens 89 mm long, 1 Acrylic Double Concave frosted lens 86mm long, 1 Universal Stainless steel mirror - can achieve the following mirrors - Plane, concave, convex - Supplied in Plastic box.