Kestrel 5000 Environmental Meter
Supplier: NIELSEN-KELLERMAN CO
0850BLK
470220-352EA 503.15 USD
470220-352
Kestrel 5000 Environmental Meter
Weather Stations
Indispensable tool for portable environmental monitoring.
  • Measures wind speed / air speed, temperature, humidity and pressure and displays over 10 total environmental parameters on large, hi-res, hi-contrast, graphic display
  • Handheld, portable environmental meter with onboard data logging and graphing
  • Protective pouch, neck lanyard and Lithium AA battery included
The Kestrel 5000 Environmental Meter is a remarkably powerful measuring and data logging tool, yet remarkably compact. The Kestrel 5000 provides the same trusted accuracy, reliability, and usability as the original Kestrel 4000. Use it for research, safety, expeditions, agriculture and any other profession or activity where accurate on-the-spot conditions monitoring is key.
Measures: Altitude, Barometric Pressure, Density Altitude, Dew Point Temperature, Heat Stress Index, Relative Humidity, Station Pressure (Absolute Pressure), Temperature, Wet Bulb Temperature (Psychrometric), Wind Chill, Wind Speed / Air Speed.