Science Take-Out® A Case Of Pesticide Poisoning
Supplier: SCIENCE TAKE-OUT, LLC
STO-147 STO-147U STO-147R STO-147-NOTG
470219-856EA 27.65 USD
470219-856 470219-858 470219-860 470219-857
Science Take-Out® A Case Of Pesticide Poisoning
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Students investigate the case of a family poisoned by household pesticides used to treat a bed bug problem.
  • Available as a Single Kit with Material for One Student or Group
  • Unassembled 10 Pack for Additional Savings
  • No Additional Equipment Needed
  • Activities Include Conducting Simulated Blood Tests and Interpreting Pesticide Labels
  • Students Explore Research on Pesticides
The investigation includes conducting simulated blood tests to identify the presence of pesticide metabolites in the family members’ blood and interpreting household pesticide labelsMore Product Information