United Scientific™ Alligator Clip Wires
Supplier: United Scientific Supplies
WAG012-PK/6 WAG024-PK/6 WAG036-PK/6 WAG012-R WAG012-B WAG024-R WAG024-B WAG036-R WAG036-B WAG012-PK/4
470219-720PK 20.8 USD
470219-720 470219-722 470219-724 470219-727 470219-729 470219-731 470219-733 470219-735 470219-737 470347-556
United Scientific™ Alligator Clip Wires
Wire
Alligator clip wires can connect to items that don't have banana plugs.
  • Protective vinyl sheath
  • Various lengths and pack sizes available
  • Flexible, 18-gauge stranded wire cord
A flexible, 18-gauge stranded wire cord has alligator clips at both ends.
Alligator clips have a protective vinyl sheath. They are available in various lengths and pack sizes.