Convection Apparatus
Supplier: United Scientific Supplies
CGA001
470219-718EA 81.1 USD
470219-718
Convection Apparatus
Models Meteorology Models
This apparatus consists of a metal box (22 x 10 x 11cm) with clear acrylic front panel, two glass chimneys, a candle, and a cotton filled wick as the smoke source. Includes activity guide.
  • Clear panel allows the flow to be seen easily
Shipping weight: 1.25 lbs.