Overhead Ripple Tank
Supplier: United Scientific Supplies
RPLTNK1
470219-716EA 3.67 USD
470219-716
Overhead Ripple Tank
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Make use of the overhead projector and shine a light on wave properties.
  • Variable speed frequecny generator
  • Includes activity manual
This device demonstrates the generation, propagation, reflection, interference, and diffraction of waves. It comes with all accessories necessary for these experiments. A 6V DC power source and an overhead projector are required (not included). Includes activity guide.
Shipping weight: 5.5 lbs. Dimensions: 15.5" x 12" x 4"