Friction Box
Supplier: United Scientific Supplies
FRBX01
470219-712EA 33.85 USD
470219-712
Friction Box
Educational Materials Physics Educational Materials Mechanics Learning Activities
Sturdy box for investigating frictional forces.
  • Smooth wood
  • Holds up to 2 kg
This friction box is used for coefficient of friction studies when larger loads are required. The 6" x 3.5" x 2.5" box will accommodate a load of 2kg or more. Smooth sanded wood with two leather strips attached to the bottom.
Shipping weight: 0.5 lbs. Dimensions: 7.5" x 4" x 3".