Volumetric Flask with Glass Stopper
Supplier: Wards
CH0442A CH0442B CH0442C CH0442E CH0442F CH0442G
470219-652EA 13 USD
470219-652 470219-654 470219-656 470219-658 470219-660 470219-662
Volumetric Flask with Glass Stopper
Flasks Volumetric Flasks
These volumetric flasks are made of borosilicate glass.
  • Flasks Fitted with Solid Glass Stoppers
  • Blue Graduation Mark
  • ASTM-E288 Compliant
  • Borosilicate Glass Construction
Each flask includes a solid glass stopper.
Each flask also has a blue graduation mark for easy measurements.
Certifications: ASTM-E288 compliant.