Spring Set
Supplier: Wards
PH0115AS
470219-544PK 135.45 USD
470219-544
Spring Set
Springs
An assortment of 150 compression and extension springs ranging from 15 to 300 mm in length, 4 to 18 mm diameters, and 16 to 32 swg in wire thickness.