Melde's Apparatus
Supplier: Wards
PH0750
470219-508EA 76.3 USD
470219-508
Melde's Apparatus
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Introduce students to different standing waves.
  • Students can even visually determine wavelength, period, and amplitude of waves
  • Unit Uses String and Oscillator to Generate Different Frequencies
For Studying the Behavior of Standing Waves