Manometer Demonstration
Supplier: Wards
PH0203G
470219-420EA 92.1 USD
470219-420
Manometer Demonstration
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Demonstrate the effect of pressure to water depth.
  • Complete package to highlight the variation of pressure in a liquid with depth
It consists of a pressure capsule of 35mm diameter placed in a bent steel tube. The pressure exerted by the water on the membrane of the capsule is measured by a pressure gauge mounted on a graduated support, connected to the probe by means of a transparent flexible tube. A device used to turn the capsule without leaving the water. The manometer tube is slideable on the support to facilitate setting of zero. Supplied with beaker.