Gas Law with Temperature Gauge
Supplier: Wards
PH0142GL
470219-418EA 138.7 USD
470219-418
Gas Law with Temperature Gauge
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
All in one unit for studying pressure, temperature and volume of a gas and to demonstrate the relationship between them.
  • Simplistic design
  • Digital temperature display with 0.1 °C resolution
  • Measures pressure from –1000 to 3000 hPa in 50hPa intervals
This unit consists of a 29 mm diameter graduated cylinder with a piston and a screw valve. The cylinder acts upon a manometer through a narrow passage.