Displacement Vessel
Supplier: Wards
PH0119C
470219-414EA 14.15 USD
470219-414
Displacement Vessel
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
A perfect tool for studying density, specific gravity, and water displacement. The 250mL polypropylene overflow can with spout only requires a catch bucket for the overflow.