Wind Turbine Generator
Supplier: Vernier Software & Technology
KW-GEN10
470219-356PK 62.99 USD
470219-356
Wind Turbine Generator
Power Supplies
This is the primary generator for wind turbine experiments because it runs smoothly and provides high power output at a relatively low RPM. This is the competition generator used in KidWind Challenge events. This generator has 4-foot leads which are attached with shrink-wrapped, reinforced soldering. Typical output in non-geared wind devices is 0.5–2.5 V at 0.1–0.3 A.