Vernier® Salinity Sensor
Supplier: Vernier Software & Technology
SAL-BTA SAL-ST
470017-558EA 156.49 USD
470017-558 470219-326
Vernier® Salinity Sensor
Electrochemistry Meters Salinity Meters
Easily and precisely measure the total dissolved salt content in ocean or brackish water.
Response time: 90% of full-scale reading in 10 s.