Digital Sensor Cable
Supplier: Vernier Software & Technology
MDC-BTD
470219-300EA 5.29 USD
470219-300
Digital Sensor Cable
Cables
This cable connects Vernier digital sensors to the digital (DIG) port on a Vernier interface.
Caution: Not compatible with the Photogate.