Vernier® AC Adapter
Supplier: Vernier Software & Technology
IPS
470219-286EA 11.99 USD
470219-286
Vernier® AC Adapter
Power Supplies
This is a 6-volt, DC, tip-negative power supply for use with Vernier LabPro, CBL 2, Digital Control Unit, and Wireless Dynamics Sensor System.